Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
CAS: 91-01-0 | C13H12O | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Catalog number FSA105391000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS13255-26-0
IUPAC Namebarium(2+) oxosilanebis(olate)
Molecular FormulaBaO3Si
InChI KeyHMOQPOVBDRFNIU-UHFFFAOYSA-N
SMILES[Ba++].[O-][Si]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)34% BaO min