1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
CAS: 89-32-7 | C10H2O6 | 218.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA105321000
Price (MYR)
343.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
343.00
Add to cart
Specifications
Chemical Name or Material1,2,4,5-Benzenetetracarboxylic anhydride
Name Note99%
CAS89-32-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger