Antipyrine, 99%, Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA104975000
Price (MYR)
607.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
607.00
Add to cart
Chemical Identifiers
CAS1067-25-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.3885-1.3925 @ 20?C