Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
CAS: 120-12-7 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA104861000
Price (MYR)
464.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Chemical Identifiers
CAS142421-57-6
IUPAC Namebis[3,5-bis(trifluoromethyl)phenyl](chloro)phosphane
Molecular FormulaC16H6ClF12P
InChI KeyDFZQEHBNAJGDCT-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(=C1)P(Cl)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White/colorless to pale yellow
FormFused or crystalline low melting solid
Assay by NMR≥96.0.% (31-P NMR)
CommentMelt may contain suspended crystals of amine hydrochloride from manufacturing process
Proton NMRConforms to structure