p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
CAS: 104-94-9 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA104830050
Price (MYR)
296.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
296.00
Add to cart
Chemical Identifiers
CAS544-17-2
IUPAC Namecalcium diformate
Molecular FormulaC2H2CaO4
InChI KeyCBOCVOKPQGJKKJ-UHFFFAOYSA-L
SMILES[Ca++].[O-]C=O.[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Complexometric Titration≥97.5 to ≤102.5%