o-Anisidine, 98+%, Thermo Scientific Chemicals
o-Anisidine, 98+%, Thermo Scientific Chemicals
o-Anisidine, 98+%, Thermo Scientific Chemicals
o-Anisidine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Anisidine, 98+%, Thermo Scientific Chemicals
CAS: 90-04-0 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA104812500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4151-80-8
IUPAC Name[4'-(dihydroxyboranyl)-[1,1'-biphenyl]-4-yl]boronic acid
Molecular FormulaC12H12B2O4
InChI KeySLHKDOGTVUCXKX-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or granular powder or lumps
Assay (Aqueous acid-base Titration)≥92.0%
Assay (HPLC)≥92.0%
Proton NMRConforms to structure