4-Aminothiophenol, 96%, Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
CAS: 1193-02-8 | C6H6NS | 124.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA104680050
Price (MYR)
368.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
368.00
Add to cart
Chemical Identifiers
CAS504-15-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to grey to light brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point105°C to 110°C
GC>=97.5 %