Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminopyrazine, 99+%, Thermo Scientific Chemicals
CAS: 5049-61-6 | C4H5N3 | 95.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA104490100
Price (MYR)
519.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
519.00
Add to cart
Specifications
Chemical Name or MaterialAminopyrazine
CAS5049-61-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.