4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Catalog number FSA104361000
Price (MYR)
354.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
354.00
Add to cart
Specifications
Chemical Name or Material4-Aminophenyl sulfone
Name Note97%
CAS80-08-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning