4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number FSA104360050
Price (MYR)
55.91
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
55.91
Add to cart
Specifications
Chemical Name or Material4-Aminophenyl sulfone
Name Note97%
CAS80-08-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning