4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenol, 97%, Thermo Scientific Chemicals
CAS: 123-30-8 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA104270050
Price (MYR)
298.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
298.00
Add to cart
Chemical Identifiers
CAS8049-11-4
IUPAC Namenonaaluminium decacopper zinc
Molecular FormulaAl9Cu10Zn
InChI KeyMPKKOQSNGWAMJD-UHFFFAOYSA-N
SMILES[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Al].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Cu].[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)44-46% Al
Assay (unspecified)4-6% Zn
Assay (unspecified)49-51% Cu