2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
CAS: 95-55-6 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA104260050
Price (MYR)
271.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
271.00
Add to cart
Chemical Identifiers
CAS51-79-6
IUPAC Nameethyl carbamate
Molecular FormulaC3H7NO2
InChI KeyJOYRKODLDBILNP-UHFFFAOYSA-N
SMILESCCOC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point46°C to 51°C
Infrared spectrumConforms
Appearance (Color)White to off-white
GC>=96.0 %