1-Aminoindan, 98%, Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Aminoindan, 98%, Thermo Scientific Chemicals
CAS: 34698-41-4 | C9H12N | 134.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA103850010
Price (MYR)
713.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
713.00
Add to cart
Chemical Identifiers
CAS4247-02-3
IUPAC Name2-methylpropyl 4-hydroxybenzoate
Molecular FormulaC11H14O3
InChI KeyXPJVKCRENWUEJH-UHFFFAOYSA-N
SMILESCC(C)COC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or lumps or flakes
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)73.0-80.0?C