2-Amino-4,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
2-Amino-4,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
2-Amino-4,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
2-Amino-4,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-4,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
CAS: 56-09-7 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA10351025025 gGlass bottle
Catalog number FSA103510250
Price (MYR)
321.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
321.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-4, 6-dihydroxypyrimidine
Name Note98%
CAS56-09-7
CAS Min %97.5
CAS Max %100.0