4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
CAS: 140-67-0 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA102880250
Price (MYR)
351.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
351.00
Add to cart
Chemical Identifiers
CAS1120-64-5
IUPAC Name2-methyl-4,5-dihydro-1,3-oxazole
Molecular FormulaC4H7NO
InChI KeyGUXJXWKCUUWCLX-UHFFFAOYSA-N
SMILESCC1=NCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Water=<0.2 % (K.F.)