4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Allylanisole, 98%, Thermo Scientific Chemicals
CAS: 140-67-0 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA102880050
Price (MYR)
225.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
225.00
Add to cart
Chemical Identifiers
CAS431-03-8
IUPAC Namebutane-2,3-dione
Molecular FormulaC4H6O2
InChI KeyQSJXEFYPDANLFS-UHFFFAOYSA-N
SMILESCC(=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.3930-1.3980 @ 20?C
Assay (GC)≥98.5%