Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
CAS: 97-59-6 | C4H6N4O3 | 158.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA102841000
Price (MYR)
351.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
351.00
Add to cart
Specifications
Chemical Name or MaterialAllantoin
Name Note98%
CAS97-59-6
CAS Min %97.5
CAS Max %100.0