Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Adipic acid, 99%, Thermo Scientific Chemicals
CAS: 124-04-9 | C6H10O4 | 146.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number FSA102810010
Price (MYR)
302.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
302.00
Add to cart
Specifications
Chemical Name or MaterialAdipic acid
CAS124-04-9
Melting Point151.0°C to 153.0°C
Boiling Point337.0°C
CAS Min %98.5