4-Acetamidophenol, 98%, Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
CAS: 103-90-2 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
5 g
250 g
1 kg
Packaging:
View all
Plastic Drum
Catalog number FSA102330051
Price (MYR)
3,481.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
3,481.00
Add to cart
Specifications
Chemical Name or Material4-Acetamidophenol
CAS103-90-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0