Microtip for use with 500 and 700 watt Sonic Dismembrators
Actual product may vary
Microtip for use with 500 and 700 watt Sonic Dismembrators
Microtips are thin, high intensity probes which are designed for processing small sample volumes. Microtips screw into the threaded end of the standard 1/2 in. probe (4220).
Have Questions?
Change view
Catalog Number
Volume Range (Metric)
Diameter (Metric) Tip
FQSFB4417
0.2 to 5 mL
2 mm
FQSFB4418
0.5 to 15 mL
3 mm
FQSFB4420
5 to 50 mL
6 mm
3 Options
Catalog number FQSFB4417
Price (MYR)
1,082.50
Add to cart
Volume Range (Metric):
0.2 to 5 mL
Diameter (Metric) Tip:
2 mm
Price (MYR)
1,082.50
Add to cart
These Fisher Scientific™ Probes are designed for use with Model 505 and Model 705 Sonic Dismembrators.
Microtip Probes for Model 505/705 Sonic Dismembrator
Requires #4220 (1/2 in. Replaceable tip probe)
Specifications
Product TypeMicrotip Probe
Amplitude320 μm maximum
Volume Range (Metric)0.2 to 5 mL
Diameter (English)0.625 in.
Diameter (English) Tip1/16 in.
Diameter (Metric)0.4 cm
Diameter (Metric) Tip2 mm
For Use With (Equipment)0.5 in. probe (#4220) for Model 505 and 705 sonic dismembrator