Check Order Status
Liquid Petroleum Gas, Specific Gravity Hydrometer with Thermometer
Actual product may vary
Liquid Petroleum Gas, Specific Gravity Hydrometer with Thermometer
The Liquid Petroleum Gas, Specific Gravity Hydrometer with Thermometer measures the SG of LPG such as propane, butane, and other light hydrocarbons.
Have Questions?
Catalog number
FKS17826583
Price (MYR)
233.13
EA
Add to cart
Price (MYR)
233.13
EA
Add to cart
Specific Gravity Range: 0.500 to 0.650
Subdivisions: 0.001
Approx. Length: 365 mm
Thermometer Range: 30 to 90°F
Thermometer Subdivision: 1°F
Thermometer filled with SafetyBLUE
Calibrated for use at 15.5°C/60°F
Specifications
Type
Liquid Petroleum Gas Hydrometer
Increments
0.001
Length (Metric)
36.5 cm
Specific Gravity Range
0.500 to 0.650
Temperature of Standardization
15°C /60°F