3 mL Graduated LDPE Pasteur Pipette
3 mL Graduated LDPE Pasteur Pipette
Actual product may vary
3 mL Graduated LDPE Pasteur Pipette
Pasteur pipette for simple and safe liquid measurement and transfer.
Have Questions?
Catalog NumberPackaging Type
FKS16404999Bulk Bag
Catalog number FKS16404999
Price (MYR)
790.00
PK
Add to cart
Packaging Type:
Bulk Bag
Price (MYR)
790.00
PK
Add to cart
  • For liquid transfer
  • Designed to ensure safe sample transfer
Specifications
AutoclavableNot Autoclavable
Bulb Draw (Metric)7 mL
CalibratedNo
Certifications/ComplianceCE Marked
Diameter (Metric) Outer7.5 mm
Dimensions (L x W)160 x 7.5 mm
MaterialLDPE
SterilityNon-sterile
Drops/mL47.6 μL
GraduatedYes
Graduation TypeMolded
Top StyleBulb
Packaging TypeBulk Bag
Volume (Metric)3 mL
Length (Metric)160 mm