Reagent Reservoir, Sterile Polystyrene
Reagent Reservoir, Sterile Polystyrene
Actual product may vary
Reagent Reservoir, Sterile Polystyrene
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)PackagingDimensions (L x W x H)
FKS16324862100 mL5/bag, 40 bags4.3 x 1.9 x 1.3 in
FKS1639485250 mL5/bag, 40 bags4 x 1.1 x 1.2 in
Catalog number FKS16324862
Price (MYR)
2,704.00
Add to cart
Capacity (Metric):
100 mL
Packaging:
5/bag, 40 bags
Dimensions (L x W x H):
4.3 x 1.9 x 1.3 in
Price (MYR)
2,704.00
Add to cart
  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents
  • Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Quality certificate available on request
  • Disposable
  • Design Patent Pending
Specifications
Capacity (English)3.38 oz
Certifications/ComplianceDnase, Rnase & Pyrogen Free
CoatingNone
ColorWhite
Description5 wrap
Capacity (Metric)100 mL
Dimensions (L x W x H)4.3 x 1.9 x 1.3 in
MaterialPolystyrene
SterilitySterile
TypeReagent Reservoir
For Use With (Application)Reagent
Height (English)1.3 in
Height (Metric)3.2 cm
IncludesReservoir
Length (English)4.3 in
Length (Metric)10.9 cm
Max. Temperature (English)104°F Operating
Max. Temperature (Metric)40°C Operating
No. per Case1200
No. per Pack200
Packaging5/bag, 40 bags
Quantity200
Volume (Metric) Well100 mL
Width (English)1.9 in
Width (Metric)4.8 cm
AutoclavableNot Autoclavable
Channels1
DisposableSingle-use
Bottom ShapeConical
Unit Size