PTFE Magnetic Stir Bar, Cross-shaped
PTFE Magnetic Stir Bar, Cross-shaped
Actual product may vary
PTFE Magnetic Stir Bar, Cross-shaped
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)
FKS1625579125 x 25 x 9mm
FKS1627579138 x 38 x 11mm
Catalog number FKS16255791
Price (MYR)
238.00
Add to cart
Dimensions (L x W x H):
25 x 25 x 9mm
Price (MYR)
238.00
Add to cart
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
  • Stable, general-purpose magnetic stirrer
  • Provides a deep, quiet vortex even at relatively high speeds
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Dimensions (L x W x H)25 x 25 x 9mm
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Height (Metric)9mm
Length (English)1 in.
Width (English)1 in.
Width (Metric)25mm
CoatingPTFE
Diameter (Metric)9 mm
Length (Metric)25 mm
MaterialPTFE
Quantity5/Pk.
ShapeCross
TypeStir Bar
Unit Size