ProCulture™ Micro-Tube Homogenizer: Pestles and Tubes
ProCulture™ Micro-Tube Homogenizer: Pestles and Tubes
Actual product may vary
ProCulture™ Micro-Tube Homogenizer: Pestles and Tubes
Chemical and heat resistant, colorant-free
Have Questions?
Catalog NumberPackaging TypeSample Volume (Metric)Product Type
FKS15795513Bulk pack1.5 mLPestle
Catalog number FKS15795513
Price (MYR)
1,741.00
PK
Add to cart
Packaging Type:
Bulk pack
Sample Volume (Metric):
1.5 mL
Product Type:
Pestle
Price (MYR)
1,741.00
PK
Add to cart
Chemical and heat resistant, colorant-free
Matched pestle tips and micro tubes can be used either with homogenizer or with hand processing.
  • Tight tolerances, thorough homogenization and grinding of materials
  • Chemical and heat resistant, colorant-free to eliminate potential leaching of heavy metals into samples
  • Manufactured and packaged in class 10,000 cleanroom
  • Certified DNase, RNase and Pyrogen-free
  • Made of animal and phalate/phalate-ester free polypropylene
  • Available in individual or bulk packs
Pestles
  • Unique ribbed shaft provides precise finger control of homogenization for shear sensitive samples
  • Fit most motorized mixer adapters
Tubes
  • Tube bottom 25% thicker bottoms than industry standard
  • Frosted side and cap surfaces for labeling, positive seal snap-cap design and pierceable top
Specifications
Product TypePestle
Voltage0/1 V
Sample Volume (Metric)1.5 mL
MaterialPolypropylene
Dnase FreeYes
For Use With (Equipment)Micro-Tube homogenizer system
Packaging Quantity100/Pk.
Packaging TypeBulk pack
RNase FreeYes
Unit SizePK