DiluCell™ 10 - Disposable Cuvette
DiluCell™ 10 - Disposable Cuvette
Actual product may vary
DiluCell™ 10 - Disposable Cuvette
Implen DiluCell™ 10 provides an automatic 1/10 dilution of higher concentrated samples for standard 1 cm cuvette spectrophotometers. Combined with low sample volume requirements and bubble free filling, the DiluCell™ allows for convenient spectrophotometric analysis from 340-1100 nm.
Have Questions?
Catalog number FKS15725805
Price (MYR)
1,919.00
EA
Add to cart
Price (MYR)
1,919.00
EA
Add to cart
Implen DiluCell™… make it virtually dilute your sample!
Previously it was necessary to use standard cuvettes with a volume of at least 1 mL and additionally to dilute the sample manually. DiluCell™ from Implen allows for lower sample volume requirements and provides automatic virtual sample dilution. Due to the unique design of DiluCell™ cuvettes, the light-path is reduced from the standard 10 mm to 0.5 mm (DC 20) or 1.0 mm (DC 10). According to the Lambert Beer Law this shortened pathlength results in an automatic sample dilution by factor 20 (DC 20) or 10 (DC 10). Automatic dilution with DiluCell™ saves time and excludes dilution errors and cross contaminations.
  • Low sample volume necessary
  • Suitable for proteins and microbial cell cultures
  • Measurement possible starting at 340 nm
  • Automatic dilution of factor 10 and factor 20
  • No risk of contamination
  • Significantly delayed cell sedimentation compared to standard cuvettes
  • Approved for usage with the Implen OD600 DiluPhotometer™
  • Made in Germany
For Research Use Only
Specifications
Description1/10 Dilution
DisposableSingle-use
Capacity (Metric)200 μL
MaterialPlastic
QuantityPack of 96
Path Length1.0 mm
Wavelength Range340 to 1100 nm
For Use With (Equipment)Spectrophotometer, OD600 DiluPhotometer
For Use With (Application)Protein assays, OD600
Diameter (Metric)12 x 12 x 45 mm
Unit SizeEA