Porcelain Plate For Desiccators
Porcelain Plate For Desiccators
Actual product may vary
Porcelain Plate For Desiccators
For use with desiccators
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Diameter (Metric) Outer
FKS15594645Desiccator (Dia.: 250mm)235 mm
FKS15584645Desiccator (Dia.: 200mm)190 mm
Catalog number FKS15594645
Price (MYR)
327.00
EA
Add to cart
For Use With (Equipment):
Desiccator (Dia.: 250mm)
Diameter (Metric) Outer:
235 mm
Price (MYR)
327.00
EA
Add to cart
  • For use with desiccators
Specifications
Diameter (Metric) Outer235 mm
Product TypePlate
For Use With (Equipment)Desiccator (Dia.: 250mm)
Unit SizeEA