Multi-Platform Shaker
Multi-Platform Shaker
Actual product may vary
Multi-Platform Shaker
Provides a smooth, quiet horizontal orbital motion for mixing in bottles, flasks and beakers
Have Questions?
Catalog NumberPlug Type
FKS15594060EU, UK, CN Plug
Catalog number FKS15594060
Price (MYR)
11,536.00
EA
Add to cart
Plug Type:
EU, UK, CN Plug
Price (MYR)
11,536.00
EA
Add to cart
  • Variable shaking speed: 50 to 450rpm 
  • Holds up to 9.9lbs, and up to 4 pieces of 140mm x 140mm sticky mat.
  • Automatic load balancing system 
  • Continuous or timed operation with automatic switch-off 
  • Maximum flexibility with many options of interchangeable accessories
  • Simple set up and easy to operate 
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Low voltage power supply provides for safe cold room operation and low energy consumption 
  • Operating temperature range of  5 to 40°C allows for use in cold rooms, refrigerators or Non-CO2 incubators
Specifications
Orbit10 mm
Load Bearing Capacity (Metric)4.5 kg
Speed Control AccuracySpeed Accuracy: ±1 rpm
Speed Range50 to 450 rpm
Voltage100/240 V
Temperature Range (Metric)5°C to 40°C
Holds250mL Flask
Load Bearing Capacity (English)9.9 lb.
Plug TypeEU, UK, CN Plug
MaterialAluminum Platform
Dimensions (LxW) Platform310 x 284 mm
Dimensions (L x W x H)415 x 293 x 120 mm
DisplayLED
IncludesAluminum Platform
No. of Positions6
Timer Range0 min. to 99:59 hr.
Unit SizeEA