CMU Series Controlled Mantle
CMU Series Controlled Mantle
Actual product may vary
CMU Series Controlled Mantle
Used in chemistry to heat liquids in round bottom flasks. Electrothermal™ CMU Series Controlled Mantle includes a stainless steel grounded earth screen covering the element and is double-fused for added safety.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (L x W x H)WattageWeight (Metric) Shipping
FKS155516201000 mL220 x 220 x 170mm380W1.5kg
Catalog number FKS15551620
Price (MYR)
8,285.00
EA
Add to cart
Capacity (Metric):
1000 mL
Dimensions (L x W x H):
220 x 220 x 170mm
Wattage:
380W
Weight (Metric) Shipping:
1.5kg
Price (MYR)
8,285.00
EA
Add to cart
  • 'Burn off' procedure at the end of the manufacturing process enables use straight out of the box.
  • Includes grounded earth screen and double fuses
  • Automatically cut out to protect the user from electric shocks.
  • Round bottom flask capacity of between 50 to 5000mL
  • Element temperatures up to 450°C
  • Built-in energy regulator
  • Indicator lamps for power and heater operation
  • Replaceable insulated heater cartridge
  • An easy to clean powder coated aluminum outer casing
  • “Cool-to-the-touch” exterior
  • Non-skid feet and support rod bracket
  • Support rodes not included
Specifications
Capacity (Metric)1000 mL
Dimensions (L x W x H)220 x 220 x 170mm
Voltage230 V
Power Supply DetailsUK and EU plug
Wattage380W
Weight (Metric) Shipping1.5kg
Temperature (Metric)450°C