Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FKS15479073 | 50 mL |
FKS15469073 | 25 mL |
FKS15489073 | 100 mL |
FKS15499073 | 150 mL |
FKS15409083 | 250 mL |
FKS15419083 | 400 mL |
FKS15429083 | 600 mL |
FKS15449083 | 1000 mL |
FKS15459083 | 2000 mL |
FKS15479083 | 5000 mL |