Check Order Status
Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Constructed of borosilicate glass. Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks are conical and graduated.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS15439103
500 mL
FKS15409103
100 mL
FKS15459103
2000 mL
FKS15419103
200 mL
FKS15429103
250 mL
FKS15449103
1000 mL
6 Options
Catalog number
FKS15439103
Price (MYR)
247.00
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
247.00
Add to cart
Erlenmeyer, graduated.
Specifications
Certifications/Compliance
ISO 1773
Diameter (Metric) Outer Neck
34 mm
Capacity (Metric)
500 mL
Material
Borosilicate Glass
Height (Metric)
180 mm
Shape
Conical
Graduated
Yes
Mouth Style
Narrow
Diameter (Metric) Outer
105 mm
Unit Size