Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Constructed from borosilicate glass. Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks are graduated and available in a variety of sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS15409113250 mL
FKS15499103100 mL
FKS15429113500 mL
Catalog number FKS15409113
Price (MYR)
252.00
Add to cart
Capacity (Metric):
250 mL
Price (MYR)
252.00
Add to cart
  • Borosilicate glass
  • Graduated
Specifications
Certifications/ComplianceDIN ISO 24450
Capacity (Metric)250 mL
Neck StyleWide neck