Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FKS15370317 | 250 mL |
More volume. More access. More space for decanting.
The new Thermo Scientific™ Nunc™ 250 mL Conical Tube offers added capacity for your larger samples with a one-of-a-kind wide mouth design, providing open, easy access for pipetting, decanting and removal of the pellet following centrifugation.