Precision™ Circulating Water Baths
Precision™ Circulating Water Baths
Actual product may vary
Precision™ Circulating Water Baths
Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionCapacity (Metric)Electrical RequirementsModel
FKS1534586735L Circulating Water Bath35 LGlobal Voltage 100/115 V 200/230 V 50/60 HzPrecision CIR 35
TSCIR3535L Circulating Water Bath35 LGlobal Voltage 100/115 V 200/230 V 50/60 HzPrecision CIR 35
Catalog number FKS15345867
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
35L Circulating Water Bath
Capacity (Metric):
35 L
Electrical Requirements:
Global Voltage 100/115 V 200/230 V 50/60 Hz
Model:
Precision CIR 35
Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.
  • Achieve enhanced temperature uniformity with perimeter-directed water flow
  • Easily clean and maintain bath with coil-free internal design
  • Optimize scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms
Not for use for IVD/MD in some countries.
Specifications
Capacity (English)9.2 gal.
Controller TypeDigital
Description35L Circulating Water Bath
Capacity (Metric)35 L
Certifications/ComplianceUL, CE
CommunicationsDigital
Temperature RangeAmbient +5° to 100°C
DisplayMonochrome LCD
Heating Capacity1500 w
Includes1 Stainless Steel Gable cover and Rubber Duck
Material304 Stainless Steel
ModelPrecision CIR 35
Temperature Presets4
Temperature Stability±0.1°C
Temperature Uniformity±0.05°C
Dimensions (D x W x H) Interior12 x 27.3 x 7.6 in. (305 x 692 x 192 mm)
Dimensions (L x W x H)15.5 x 36.9 x 9.8 in. (394 x 938 x 249 mm)
Electrical RequirementsGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Amperage11.2-13.1 / 5.4-6.3 (120V/230V)
Unit Size