Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Actual product may vary
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Ideal for work with narrow tubes and containers with small openings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityBulb Draw (Metric)Packaging Type
FKS153178431.3 mLNon-sterile0.8 mLBulk
FKS153878434.5 mLNon-sterile3.3 mLBulk
6946.9 mLNon-sterile3.2 mLBulk
Catalog number FKS15317843
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1.3 mL
Sterility:
Non-sterile
Bulb Draw (Metric):
0.8 mL
Packaging Type:
Bulk
Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Narrow Stem Transfer Pipettes. These polyethylene pipettes prevent cross-contamination and are used narrow tubes and containers with small openings . The single-unit design features a built-in pipette bulb, and a flexible stem bends to sample from narrow or small tubes.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)0.8 mL
DescriptionTransfer Pipette, SEDI-PET, Short Stem, 1.3mL
Diameter (Metric) Outer2.5 mm
MaterialLDPE
SterilityNon-sterile
Drops/mL26
GraduatedNo
Quantity500/Pk 8Pk/Cs 4000/Cs
Packaging TypeBulk
Volume (Metric)1.3 mL
Length (Metric)6.4 cm
TypeNarrow Stem Transfer Pipet