For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Have Questions?
Catalog number FKS15302005
Price (MYR)
246.00
PK
Add to cart
Price (MYR)
246.00
PK
Add to cart
Calcium Hardness Tablets
Method: Tablet Count
Bottle of 250
Specifications
For Use With (Application)Calcium Hardness Test
For Use With (Equipment)Test kits, colorimeters and spectrophotometers