DURAN™ Crystallizing Dish, without spout
DURAN™ Crystallizing Dish, without spout
Actual product may vary
DURAN™ Crystallizing Dish, without spout
DURAN™ Crystallizing Dish, without spout
DURAN™ Crystallizing Dish, without spout
DURAN™ Crystallizing Dish, without spout
DURAN™ Crystallizing Dish, without spout
The flat shape as well as the flat bottom of the crystallizing dish allow an even evaporation of solvents and thus the crystallization of solids from solutions and suspensions.
Have Questions?
Catalog NumberCapacity (Metric)
FKS152216893500 mL
Catalog number FKS15221689
Price (MYR)
12,500.00
PK
Add to cart
Capacity (Metric):
3500 mL
Price (MYR)
12,500.00
PK
Add to cart
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
  • Flat and wide shape allow solvents to evaporate quickly and evenly
  • Large ground base
  • Spout for clean pouring
  • Fired-on, highly durable white ceramic
Specifications
AutoclavableAutoclavable
Capacity (Metric)3500 mL
MaterialBorosilicate Glass (3.3)
For Use With (Application)Crystallization; Preparation of concentrated solutions; Available with spout DIN 12 338 or without spout DIN 12 337
No. per Pack10/Pk.
Diameter (Metric)230 mm
Unit SizePK