Stainless Steel Jug Graduated with Hinged Lid
Stainless Steel Jug Graduated with Hinged Lid
Actual product may vary
Fisherbrand
Stainless Steel Jug Graduated with Hinged Lid
Get optimium cleaning and sterilizing results with less effort. Suitable for pharmaceutical and laboratory applications, Fisherbrand™ Stainless Steel Jug Graduated with Hinged Lid, is the material of choice for applications requiring sterile surfaces. Pour easier with this durable, spouted, 304 spec stainless steel seamless jug. All surfaces are non-reacting, highly polished, and offer zero absorption making cleaning easier.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS15218846500 mL
FKS152688462000 mL
Catalog number FKS15218846
Price (MYR)
461.00
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
461.00
Add to cart
  • Material of choice for applications requiring sterile surfaces
  • Suitable for pharmaceutical and laboratory applications  
  • Spouted for easy pouring 
  • Internal seamless design with a beaded rim for durability 
  • 304 spec stainless steel seamless Jugs
  • All surfaces are non-reacting, highly polished, and offer zero absorption making cleaning easier
  • Immune to sterilizing solutions, most of the organic chemicals and dyestuffs, and a wide variety of inorganic chemicals.
Specifications
Diameter (Metric) Outer90 mm
Capacity (Metric)500 mL
MaterialStainless Steel
Height (Metric)135 mm
IncludesHinged Lid
TypeJug