Replacement Stopcocks
Replacement Stopcocks
Actual product may vary
Replacement Stopcocks
Polypropylene body has Teflon™ PTFE plug, washer and bright red handle
Have Questions?
Catalog number FKS15109034
Price (MYR)
973.00
EA
Add to cart
Price (MYR)
973.00
EA
Add to cart
Polypropylene body has Teflon™ PTFE plug, washer and bright red handle
  • Three-way plug for vacuum draw, shut-off and vacuum release
  • Taper fits on desiccator porte
Use on Desiccator Models: 08-594-15A, 08-594-16A, 08-594-15B, 08-594-16B, 08-594-15C, 08-594-16C, 01-817-50
Teflon™ is a registered trademark of the Chemours Company FC LLC used under license by Bel-Arts Products, Inc.
Specifications
Type3-way Stopcock
Plug Bore0.24 in. x 0.38 in. O.D.
MaterialPolypropylene
For Use With (Application)Vacuum Desiccator Cabinets
For Use With (Equipment)Scienceware™ desiccators, space saver and Techni-Dome™ 360 models
IncludesWasher, high visibility bright red handle with nut
Plug MaterialPTFE
Outlet PortMale Slip
Connectors3-way
Unit SizeEA