High-Form Porcelain Crucibles
High-Form Porcelain Crucibles
Actual product may vary
High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Thickness (Metric)Diameter (Metric) Outer
FKS1344969930 mL37mm2mm43mm
FISFB965E30 mL37mm2mm43mm
FISFB965G50 mL43mm2.1mm53mm
FKS1348969950 mL43mm2.1mm53mm
FKS13499699100 mL54mm2.1mm65mm
FISFB965F40 mL40mm2 mm50mm
FKS1340970940 mL40mm2 mm50mm
Catalog number FKS13449699
Price (MYR)
414.00
PK
Add to cart
Capacity (Metric):
30 mL
Height (Metric):
37mm
Thickness (Metric):
2mm
Diameter (Metric) Outer:
43mm
Price (MYR)
414.00
PK
Add to cart
Withstand temperatures up to 1150°C (2102°F)
  • Glazed inside and out except for outside bottom surface and rim
Specifications
MaterialPorcelain
FinishGlazed
Max. Temperature (Metric)1150°C
Thickness (Metric)2mm
Weight (Metric)26g
Capacity (Metric)30 mL
Diameter (Metric) Outer43mm
Height (Metric)37mm
Product TypeCrucible
Unit SizePK