Porcelain Evaporating Dishes
Porcelain Evaporating Dishes
Actual product may vary
Porcelain Evaporating Dishes
Glazed inside and out except for rim.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberdimCapacityMetric
FKS13439729
FKS13499719
FKS13419729
FISFB968F
FKS13429729
FISFB962M
Catalog number FKS13439729
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Porcelain dish with lip
  • Glazed inside and out except for rim
  • Highly resistant to chemicals
  • Withstand temperatures up to 1150°C (2102°F)
  • Without cover
Specifications
Diameter (English)4.52 in.
Unit Size