Non-sterile Transfer Pipettes, Transparent LDPE
Non-sterile Transfer Pipettes, Transparent LDPE
Actual product may vary
Fisher Catalogue
Non-sterile Transfer Pipettes, Transparent LDPE
Prevent cross contamination. Fisherbrand™ Non-sterile Transfer Pipets, Transparent LDPE are made of shatterproof, flexible plastic and are designed to replace glass Pasteur pipets. Graduated and non-graduated pipets available in a range of sizes and styles, including jumbo, thin stem and extra long.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Drops/mLGraduatedTip Style
FKS134391083 mL25YesStandard
FKS134491087 mL20NoExtra Long
Catalog number FKS13439108
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
3 mL
Drops/mL:
25
Graduated:
Yes
Tip Style:
Standard
  • Built-in bulb eliminates the need for plugging, packaging and autoclaving
  • Flexible stem bends to sample from narrow or small tubes
  • Low affinity surface reduces loss of cells and valuable proteins due to binding
  • Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
  • Non-sterile, transparent and fully recyclable
Specifications
MaterialLDPE
Drops/mL25
GraduatedYes
Graduations0.5 mL
Quantity500 Pack
Tip StyleStandard
Volume (Metric) Dispense40 μL drop
Volume (Metric)3 mL
Length (Metric)155 mm
TypeTransfer Pipet
Unit Size