High-Form Porcelain Crucibles
High-Form Porcelain Crucibles
Actual product may vary
High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Thickness (Metric)Diameter (Metric) Outer
FKS1340970940 mL40mm2 mm50mm
FISFB965E30 mL37mm2mm43mm
FKS1344969930 mL37mm2mm43mm
FISFB965G50 mL43mm2.1mm53mm
FKS1348969950 mL43mm2.1mm53mm
FKS13499699100 mL54mm2.1mm65mm
FISFB965F40 mL40mm2 mm50mm
Catalog number FKS13409709
Price (MYR)
426.00
Add to cart
Capacity (Metric):
40 mL
Height (Metric):
40mm
Thickness (Metric):
2 mm
Diameter (Metric) Outer:
50mm
Price (MYR)
426.00
Add to cart
Withstand temperatures up to 1150°C (2102°F)
  • Glazed inside and out except for outside bottom surface and rim
Specifications
MaterialPorcelain
FinishGlazed
Max. Temperature (Metric)1150°C
Thickness (Metric)2 mm
Weight (Metric)32g
Capacity (Metric)40 mL
Diameter (Metric) Outer50mm
Height (Metric)40mm
Product TypeCrucible
Unit Size