LO-tox™ Lab Thermometer
LO-tox™ Lab Thermometer
Actual product may vary
Fisherbrand
LO-tox™ Lab Thermometer
LO-tox™ lab thermometer without calibration certificate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)SubdivisionImmersion TypeLength (Metric)
FKS13058603-10°C to +101°C0.2°CPartial Immersion405 mm
FKS15432695-10°C to +360°C
Catalog number FKS13058603
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
-10°C to +101°C
Subdivision:
0.2°C
Immersion Type:
Partial Immersion
Length (Metric):
405 mm
Read temperatures quickly and accurately with blue spirit against a white background. Brannan™ Lo-Tox™ Spirit-filled partial Immersion Thermometer is manufactured to traceable standards and includes a blue non-roll top and safety suspension ring.
  • General purpose/stirring thermometer with a low toxic, environmentally friendly filling
  • Easy to read against background
  • Each thermometer is manufactured to traceable standards
Specifications
Bottle FillBlue Spirit
For Use With (Application)General use
Immersion TypePartial Immersion
IncludesBrannan non roll end cap
Length (Metric)405 mm
MaterialGlass
Product TypeLaboratory Thermometer
Max. Temperature (Metric)101°C
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +101°C
Accuracy±1°C
Subdivision0.2°C
Immersion Length76 mm
Unit Size