Borosilicate Glass Automatic Burette
Borosilicate Glass Automatic Burette
Actual product may vary
Fisherbrand
Borosilicate Glass Automatic Burette
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Product Type
FKS1288209450 mLAutomatic Buret
FKS1289209410 mLAutomatic buret w/ Schellbach stripe
Catalog number FKS12882094
Price (MYR)
2,678.00
Add to cart
Volume (Metric):
50 mL
Product Type:
Automatic Buret
Price (MYR)
2,678.00
Add to cart
  • With automatic zero facility
  • Simple operation - To fill burette apply a slight positive pressure to lower sidearm - To set zero, release pressure to allow excess liquid to siphon back into the bottle
  • Burettes manufactured from chemically resistant Pyrex™ borosilicate glass
  • White enamel graduations
  • 1.5mm bore PTFE key stopcock
  • The following accessories are sold separately: Glass Reservoir (3297/12) and Rubber Blowing Ball for Class AS Auto Burets (3297/14AS)
Specifications
Product TypeAutomatic Buret
Certifications/ComplianceISO 385 and DIN 12700
MaterialChemically Resistant Pyrex Borosilicate Glass
Volume (Metric)50 mL
Includes1.5 mm bore PTFE key stopcock
IncrementsWhite enamel graduations at 0.10 mL
Unit Size