Test Tube Brushes
Test Tube Brushes
Actual product may vary
Fisherbrand
Test Tube Brushes
Excellent for cleaning out narrow test tubes and other hard-to-reach instruments. Pinnacle Brush™ Test Tube Brushes are supplied in packs of 10.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBristle MaterialDiameter (Metric) BristleFor Use With (Equipment)
FKS12871954Fiber29 mm16 to 25 mm dia. Test Tubes
FKS12861954Fiber11 mm10 to 12 mm dia. Test Tubes
FKS12891954Nylon28 mm16 to 25 mm dia. Test Tubes
Catalog number FKS12871954
Price (MYR)
183.00
Add to cart
Bristle Material:
Fiber
Diameter (Metric) Bristle:
29 mm
For Use With (Equipment):
16 to 25 mm dia. Test Tubes
Price (MYR)
183.00
Add to cart
Excellent for cleaning out narrow test tubes and other hard-to-reach instruments. Pinnacle Brush™ Test Tube Brushes are supplied in packs of 10.
  • On galvanized steel wire handles
  • With fan-shaped end
  • With bristle or nylon tips
  • Available for different diameters of tubes
Specifications
Product TypeTest Tube Brush
Bristle MaterialFiber
Handle MaterialMetal
Tip TypeFan Tip
ColorWhite
Diameter (Metric)3.2 cm
Diameter (Metric) Bristle29 mm
For Use With (Equipment)16 to 25 mm dia. Test Tubes
Length (Metric)22 cm
Length (Metric) Brush6 cm
Length (Metric) Handle16 cm
Quantity10 Pk.
SizeBrush Circumference: 10 cm
Unit Size