M-Zone™ Bottle Brush
M-Zone™ Bottle Brush
Actual product may vary
M-Zone™ Bottle Brush
Color-coded bottle brushes with rounded, stiff polyester bristles for versatile and aggressive cleaning of bottles and various equipment in production and in the lab.
Have Questions?
Catalog NumberColorDiameter (Metric) Bristle
FKS12765755Yellow8.26 cm
Catalog number FKS12765755
Price (MYR)
499.00
EA
Add to cart
Color:
Yellow
Diameter (Metric) Bristle:
8.26 cm
Price (MYR)
499.00
EA
Add to cart
A range of color-coded bottle brushes with rounded, stiff polyester bristles for versatile and aggressive cleaning of bottles and various equipment in production and in the lab.
The Micronova M-Zone™ Bottle Brushes are multipurpose brushes with rounded, stiff polyester bristles and either 12 or 16-inch plastic easy grip handles. Designed to clean bottles and other lab and production equipment.
  • Multipurpose, color-coded plastic bottle brushes
  • Durable, stiff polyester bristles
  • Easy grip plastic handle
  • For versatile and aggressive cleaning of bottles and equipment
  • Made from FDA approved, BPA free base resins
  • Available in Blue, Green, Red, White and Yellow
Specifications
Product TypeBottle Brush
Bristle MaterialPolyester
Bristle TypeSoft
Color-codedYes
Handle MaterialPolypropylene
Handle TypeRound
Tip TypeCurved
SterilityNon-sterile
AppearanceRounded, yellow, Bottle Brush Head with soft bristles, attached to a fixed length, yellow, plastic handle with rounded end.
AutoclavableNot Autoclavable
Care InstructionsKeep at ambient temperature
Certifications/ComplianceCertificate of Conformance; ISO 9001 Certified Facility; FDA Approved
ColorYellow
Diameter (English) Bristle3.25 in.
Diameter (Metric) Bristle8.26 cm
Dimensions (L x W x H)16 x 3.5 x 3.5 in (40.6 x 8.89 x 8.89 cm)
DisposableReusable
FilteredNon-filtered
FlammableNo
For Use With (Application)Fine cleaning of bottles and various equipment in labs, cleanrooms and production facilities.
For Use With (Equipment)BRSC6-01: NovaPoly Brush Cover; BRSC68-01: PolyMesh Brush Cover
Height (English)3.5 in.
Height (Metric)8.89 cm
IncludesHandle/Included, Part of Brush
Length (English)16 in.
Length (English) Bristle3.25 in.
Length (English) Brush6.5 in.
Length (English) Handle9.75 in.
Length (Metric)40.6 cm
Length (Metric) Bristle8.26 cm
Length (Metric) Brush16.5 cm
Length (Metric) Handle24.77 cm
Length (Metric) Tip6.35 mm
MaterialPolypropylene, Polyester
No. of Bristle Rows19
PackagingEach brush single bagged
Quantity12/Cs.
Resistant ToStandard disinfectants and cleaning solutions
Size16 x 10 x 6 in.
Width (English)3.5 in.
Width (Metric)8.89 cm
Width (English) Bristle3.25 in.
Width (Metric) Bristle8.26 cm
Unit SizeEA
Contents & Storage
Keep at ambient temperature