Polypropylene Pop-Up Tube Racks
Polypropylene Pop-Up Tube Racks
Actual product may vary
Fisherbrand
Polypropylene Pop-Up Tube Racks
Opens and locks in place easily for quick use. Polypropylene Pop-Up Tube Racks provide space saving and compact storage for 15 and/or 50 mL tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHolds
FKS12603476GreenTubes (50 mL)
FKS12693466PurpleTubes (15 mL), Tubes (17 mm)
Catalog number FKS12603476
Price (MYR)
469.00
Add to cart
Color:
Green
Holds:
Tubes (50 mL)
Price (MYR)
469.00
Add to cart
  • Made from polypropylene
  • Easy to open – easy to close
  • Designed to help organize 15 and 50 mL tubes
  • Imprinted grid aids quick retrieval
  • Stack racks in open or closed position
  • Saves space when racks are not in use
  • Not recommended for cleaning in autoclave
Specifications
IncludesImprinted Grid
Product TypeCollapsible Tube Rack
AutoclavableNot Autoclavable
ColorGreen
HoldsTubes (50 mL)
MaterialPolypropylene
Positions18
ShapeRectangular
ShelvesNone
StackableYes
SterilityNon-sterile
For Use With (Equipment)50 mL Test Tubes
Quantity2 Pk.
Dimensions (L x W x H)255 x 137 x 72 mm
Length (Metric)255 mm
Length (English)10 in.
Width (Metric)137 mm
Height (Metric)72 mm
Height (English)2.8 in.
Diameter (Metric) Opening30 mm
Weight (Metric)1 kg
Unit Size