Azlon™ Multi-lingual Safety Venting Wash Bottle
Azlon™ Multi-lingual Safety Venting Wash Bottle
Actual product may vary
Azlon™ Multi-lingual Safety Venting Wash Bottle
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Chemical LabelDescriptionClosure Color
FKS12357512500 mLEthanolRegion 2: Norwegian, Danish, Swedish, EnglishOrange
FKS12196220250 mLIsopropanolPrinted in English, French, German and SpanishBlue
FKS12550456500 mLMethanolPrinted in English, French, German and SpanishGreen
FKS11303524500 mLIsopropanolPrinted in English, French, German and SpanishBlue
Catalog number FKS12357512
Price (MYR)
500.00
PK
Add to cart
Capacity (Metric):
500 mL
Chemical Label:
Ethanol
Description:
Region 2: Norwegian, Danish, Swedish, English
Closure Color:
Orange
Price (MYR)
500.00
PK
Add to cart
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
  • Multi-lingual wide neck wash bottles featuring the patented DripLok™ vapor-venting valve that prevents solvent drips
  • Indelibly printed in multiple languages (based on region)
  • Includes the Globally Harmonised Symbols, NFPA diamond, risk phrases, and safety information
  • Fitted with color coded one piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)500 mL
DescriptionRegion 2: Norwegian, Danish, Swedish, English
MaterialLDPE
Closure ColorOrange
Height (Metric)163 mm
Chemical LabelEthanol
Diameter (Metric)75 mm
Unit SizePK