Tall Form Porcelain Crucibles Without Lid
Tall Form Porcelain Crucibles Without Lid
Actual product may vary
Tall Form Porcelain Crucibles Without Lid
Use these dishes for a variety of applications. Haldenwanger™ Tall Form Porcelain Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1230651740 mL
FKS1239650724 mL
Catalog number FKS12306517
Price (MYR)
219.96
PK
Add to cart
Capacity (Metric):
40 mL
Price (MYR)
219.96
PK
Add to cart
  • Available in a variety of sizes
  • Sizes up to 175mL glazed except rim
  • 1,000mL and larger are glazed except outside base
  • Complies with DIN VDE 0335, type C 100
Specifications
MaterialPorcelain
FinishGlazed
Capacity (Metric)40 mL
Diameter (Metric) Outer40mm
Height (Metric)50mm
Product TypeTall Form Porcelain Crucibles
Unit SizePK